2-Methoxy-5-(trifluoromethyl)phenylboronic acid ≥98%
Warning
Synonyms: 2-Methoxy-5-(trifluoromethyl)benzeneboronic acid
H52932.MDEA 36.5 EUR
H52932.MD H52932.03
2-Methoxy-5-(trifluoromethyl)phenylboronic acid ≥98%
2-Methoxy-5-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219.95
Melting Pt: 144…148 °C
Storage Temperature: Ambient
MDL Number: MFCD04039222
CAS Number: 240139-82-6