Furfuryl methyl disulphide for synthesis
Supplier: Merck
Synonyms: Methyl furfuryl disulfide
M-Clarity™ quality level = MQ200
8.43860.0025EA 179 EUR
8.43860.0025 8.43860.0005
Furfuryl methyl disulphide for synthesis
Furfuryl methyl disulphide
Formula: C₆H₈OS₂
Storage Temperature: Ambient
MDL Number: MFCD00012326
CAS Number: 57500-00-2
EINECS: 260-773-7