2-Mercapto-6-nitrobenzothiazole ≥96%
Supplier: Thermo Scientific
Warning
Synonyms: 6-Nitrobenzo[d]thiazole-2(3H)-thione
A18211.06EA 203 EUR
A18211.06 A18211.03
2-Mercapto-6-nitrobenzothiazole ≥96%
2-Mercapto-6-nitrobenzothiazole
Formula: C₇H₄N₂O₂S₂
MW: 212.24
Melting Pt: 250…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00041850
CAS Number: 4845-58-3