3-Carboxyphenylboronic acid
Supplier: Thermo Scientific
Warning
Synonyms: 3-Carboxybenzeneboronic acid3-Boronobenzoic acid
B25315.06EA 141 EUR
B25315.06 B25315.03
3-Carboxyphenylboronic acid
3-Carboxyphenylboronic acid
Formula: C₇H₇BO₄
MW: 165.94
Melting Pt: ∼ 240 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00036833
CAS Number: 25487-66-5
EINECS: 000-000-0