2-Chloro-5-(2,4-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
H54815.MDEA 61.5 EUR
ALFAH54815.MD ALFAH54815.03
2-Chloro-5-(2,4-dimethylphenylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BClNO3
Formula Weight: 303.55
MDL No.: MFCD22682830