Imprimer
4-Chloro-2-(ethoxycarbonyl)benzeneboronic acid, 95%
H53153.03EA 182 EUR
ALFAH53153.03 ALFAH53153.MD
4-Chloro-2-(ethoxycarbonyl)benzeneboronic acid, 95%
Purity: 95%
CAS No.: 850568-61-5
Molecular Formula: C9H10BClO4
Formula Weight: 228.44
Melting Point: 132-134°
MDL No.: MFCD06659823