5-Carboxy-2-chlorobenzeneboronic acid, 98%
H52583.03EA 53.3 EUR
ALFAH52583.03 ALFAH52583.MD
5-Carboxy-2-chlorobenzeneboronic acid, 98%
Purity: 98%
CAS No.: 913835-75-3
Molecular Formula: C7H6BClO4
Formula Weight: 200.39
Melting Point: 228-244°
MDL No.: MFCD08689491