3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 95%
Supplier: Thermo Scientific
Warning
Synonyms: 3-(Dimethylcarbamoyl)-2-Fluorobenzeneboronic Acid
H56087.MDEA 190 EUR
H56087.MD H56087.03
3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid 95%
3-(Dimethylcarbamoyl)-2-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD16295256
CAS Number: 1451391-88-0