4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: 4-Dimethylcarbamoyl-2-fluorobenzeneboronic acid
H55993.03EA 760 EUR
H55993.03 H55993.MD
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid ≥97%
4-(Dimethylcarbamoyl)-2-fluorophenylboronic acid
Formula: C₉H₁₁BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD11855833
CAS Number: 874289-30-2