3-(2,5-Dimethylphenylcarbamoyl)-5-fluorobenzeneboronic acid, 97%

H54591.MDEA 178 EUR
ALFAH54591.MD ALFAH54591.03
3-(2,5-Dimethylphenylcarbamoyl)-5-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040225
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR