5-(2,4-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%

H54260.MDEA 86 EUR
ALFAH54260.MD ALFAH54260.03
5-(2,4-Dimethylphenylcarbamoyl)-2-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.09
MDL No.: MFCD20040164
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR