3-Ethoxycarbonyl-4-fluorobenzeneboronic acid, 98%

H52528.03EA 359 EUR
ALFAH52528.03 ALFAH52528.MD
3-Ethoxycarbonyl-4-fluorobenzeneboronic acid, 98%
Purity: 98%
CAS No.: 874219-36-0
Molecular Formula: C9H10BFO4
Formula Weight: 211.98
Melting Point: 149-152°
MDL No.: MFCD08061836
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR