3-Methyl-5-(trifluoromethoxy)benzylamine 97%
Supplier: Alfa Aesar
Danger
H31840.03EA 173 EUR
H31840.03 H31840.MD
3-Methyl-5-(trifluoromethoxy)benzylamine 97%
3-Methyl-5-(trifluoromethoxy)benzylamine
Formula: C₉H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09025379
CAS Number: 916420-60-5
ADR: 8,III