2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone ≥97%
Supplier: Thermo Scientific
Danger
Synonyms: 3,5-Bis(tert-butyl)-4-hydroxyphenacyl bromide
H26835.06EA 200 EUR
H26835.06 H26835.14 H26835.03
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone ≥97%
2-Bromo-3',5'-di-tert-butyl-4'-hydroxyacetophenone
Formula: C₁₆H₂₃BrO₂
MW: 327.26
Melting Pt: 104…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00173889
CAS Number: 14386-64-2
UN: 1759
ADR: 8,III