Imprimer
3-[Benzyl(ethyl)carbamoyl]-5-fluorobenzeneboronic acid, 97%
H54376.03EA 344 EUR
ALFAH54376.03 ALFAH54376.MD
3-[Benzyl(ethyl)carbamoyl]-5-fluorobenzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C16H17BFNO3
Formula Weight: 301.12
MDL No.: MFCD20040177