Imprimer
3-Fluoro-4-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
H54326.03EA 344 EUR
ALFAH54326.03 ALFAH54326.MD
3-Fluoro-4-(4-methylbenzylcarbamoyl)benzeneboronic acid, 97%
Purity: 97%
Molecular Formula: C15H15BFNO3
Formula Weight: 287.10
MDL No.: MFCD20265242