Nickel(II) chloride 0.2 mol/l in aqueous solution

Supplier: CUSTOM MADE CHEMICALS LAB

Synonyms: Nickel dichloride

CMPLH005.W.2.5L.40EA 0 EUR
CMPLH005.W.2.5L.40
Nickel(II) chloride 0.2 mol/l in aqueous solution
Nickel (II) chloride
Formula: Cl₂Ni
MW: 129.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3287
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR