3-Fluoro-5-methylphenylboronic acid ≥98%
Warning
Synonyms: 3-Fluoro-5-methylbenzeneboronic acid
H53111.03EA 188 EUR
H53111.03 H53111.MD
3-Fluoro-5-methylphenylboronic acid ≥98%
3-Fluoro-5-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95
Melting Pt: 216…222 °C
Storage Temperature: Ambient
MDL Number: MFCD07363785
CAS Number: 850593-06-5