2'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97%
Supplier: Thermo Scientific
Warning
B22931.06EA 197 EUR
B22931.06 B22931.03
2'-Fluoro-3'-(trifluoromethyl)acetophenone ≥97%
2'-Fluoro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
Density: 1.020
Storage Temperature: Ambient
MDL Number: MFCD00061145
CAS Number: 207853-63-2