3,5-Difluoro-2-methoxybenzylamine 97%
Supplier: Thermo Scientific
Danger
H26521.03EA 145 EUR
H26521.03 H26521.MD
3,5-Difluoro-2-methoxybenzylamine 97%
3,5-Difluoro-2-methoxybenzylamine
Formula: C₈H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD06660178
CAS Number: 886502-00-7
ADR: 8,III