2-Methyl-3-(trifluoromethyl)benzyl bromide ≥97%
Supplier: Thermo Scientific
Danger
H26280.03EA 167 EUR
H26280.03 H26280.MD
2-Methyl-3-(trifluoromethyl)benzyl bromide ≥97%
2-Methyl-3-(trifluoromethyl)benzyl bromide
Formula: C₉H₈BrF₃
Boiling Pt: 72 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01631608
CAS Number: 261952-16-3
EINECS: None
ADR: 8,III