2',4'-Difluoro-3'-methoxyacetophenone ≥97%
Supplier: Thermo Scientific
Warning
H26456.06EA 308 EUR
H26456.06 H26456.03
2',4'-Difluoro-3'-methoxyacetophenone ≥97%
2',4'-Difluoro-3'-methoxyacetophenone
Formula: C₉H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02258906
CAS Number: 373603-19-1