Chloroform/n-Heptane/Methanol mixture (49:49:2)% (v/v/v)
Danger
PRLSMC311776.5000EA 0 EUR
PRLSMC311776.5000 PRLSMC311776.2500
Chloroform/n-Heptane/Methanol mixture (49:49:2)% (v/v/v)
Mixture of chloroform/n-heptane/methanol (49:49:2 %vol.)
Storage Temperature: Ambient
UN: 1992
ADR: 3,II