Mixture of alcohols

Danger

PRLS89249.320EA 0 EUR
PRLS89249.320
Mixture of alcohols
Mixture of alcohols
Storage Temperature: Ambient
UN: 2920
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR