2'-Chloro-6'-fluoro-3'-methylacetophenone ≥97%
Supplier: Thermo Scientific
Warning
B24003.06EA 213 EUR
B24003.06 B24003.03
2'-Chloro-6'-fluoro-3'-methylacetophenone ≥97%
2'-Chloro-6'-fluoro-3'-methylacetophenone
Formula: C₉H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD01631368
CAS Number: 261762-63-4