4-Bromo-2-ethylaniline hydrochloride ≥98%
Danger
A13077.14EA 123 EUR
A13077.14 A13077.06
4-Bromo-2-ethylaniline hydrochloride ≥98%
4-Bromo-2-ethylaniline hydrochloride
Formula: C₈H₁₁BrClN
Melting Pt: ∼ 222 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00156127
CAS Number: 30273-22-4