2-Methoxyethyl phenyl sulphide ≥98%
Supplier: Thermo Scientific
L10464.14EA 134 EUR
L10464.14 L10464.06
2-Methoxyethyl phenyl sulphide ≥98%
2-Methoxyethyl phenyl sulphide
Formula: C₉H₁₂OS
Boiling Pt: 125…126 °C (18 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00041329
CAS Number: 77298-24-9