(1-Ethylpropyl)benzene ≥99.0% (by GC)

Supplier: TCI EUROPE
Warning

TCIAE0305-1MLEA 189 EUR
TCIAE0305-1ML
(1-Ethylpropyl)benzene ≥99.0% (by GC)
(1-Ethylpropyl)benzene
Formula: C₁₁H₁₆
Boiling Pt: 191°C
MDL Number: MFCD00048663
CAS Number: 1196-58-3
UN: 3295
ADR: 3,III
https://be.vwr.com/stibo/web/std.lang.all/78/43/28967843.jpg

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR