Taurine Cell Culture Reagent

Supplier: Thermo Scientific

Synonyms: 2-Aminoethanesulphonic acid

J62308.A1EA 427 EUR
J62308.A1 J62308.22
Taurine Cell Culture Reagent
Taurine
Formula: C₂H₇NO₃S
MW: 125.15 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00008197
CAS Number: 107-35-7
EINECS: 203-483-8
Merck Index: 12,09241

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR