3,4-Difluorotoluene ≥98.0% (by GC)
Supplier: TCI EUROPE
|
Warning
|
TCIAD4054-5GEA
105
EUR
TCIAD4054-5G
TCIAD4054-1G
3,4-Difluorotoluene ≥98.0% (by GC)
3,4-Difluorotoluene
Formula:
C₇H₆F₂ MW: 128.12 g/mol Boiling Pt: 112°C Flash Pt: 25°C |
MDL Number:
MFCD00075087 CAS Number: 2927-34-6 UN: 1993 ADR: 3,III |
![]() |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |