5-Chloro-2,4-dimethoxyaniline ≥98%
Warning
H59214.03EA 42.9 EUR
H59214.03
5-Chloro-2,4-dimethoxyaniline ≥98%
5-Chloro-2,4-dimethoxyaniline
Formula: C₈H₁₀ClNO₂
MW: 187.63 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 87…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00025752
CAS Number: 97-50-7
EINECS: 202-586-5
UN: 3077
ADR: 9,III