Caprylic acid, Supelco®
Supplier: Merck
Danger
Synonyms: n-Octanoic AcidOctoic acidOctanoic acid
PHR1202-1GEA 83.5 EUR
PHR1202-1G
Caprylic acid, Supelco®
Caprylic acid
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 239.7 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.9066 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00004429
CAS Number: 124-07-2
EINECS: 204-677-5
UN: 3265
ADR: 8,III
Merck Index: 13,01771