Ammonium chloride 0.5 M in aqueous solution
Supplier: Thermo Scientific
J62046.ADEA 33.9 EUR
J62046.AD J62046.AE
Ammonium chloride 0.5 M in aqueous solution
Ammonium chloride
Formula: NH₄Cl
MW: 53.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
https://be.vwr.com/stibo/web/std.lang.all/05/71/16640571.jpg