Electrophoresis systems, PhastSystem™
Supplier: Cytiva
18-1200-10EA 13055 EUR
GEHE18-1200-10
Electrophoresis systems, PhastSystem™
PhastSystem™ is a multi-tasking, semi-automated electrophoresis workstation that runs up to two gels, under the same conditions within a separation and control unit, while staining up to two gels in a development unit.
  • Automatically separate and stain proteins under controlled power and temperature conditions
  • Program and store nine separation protocols of nine steps each, and nine staining protocols of 20 steps each
  • Regulate gel temperature through Peltier-controlled support bed
  • Assure reproducibility with quality tested precast gels, buffer strips and staining kits
  • Short circuit protection; protected against power disturbances and static discharges; lid power interlock and alarm