NHS HP SpinTrap™ Buffer Kit
Supplier: Cytiva
28-9135-69EA 323 EUR
28-9135-69
NHS HP SpinTrap™ Buffer Kit
Buffers Biological Buffers
NHS HP SpinTrap™ Buffer kit is optimised for standard affinity-based enrichment/immunoprecipitation according to the protocol provided with the NHS HP SpinTrap™ columns.
  • High quality buffers
  • All buffer components listed and defined
  • Convenient format - stock solutions can be diluted directly into the provided bottles for storage
  • Eliminates time-consuming buffer preparation, which allows fast, reproducible protein enrichment