Buffer, AMPSO, 0,2M solution, pH 8,0

J61467.AKEA 44.7 EUR
ALFAJ61467.AK ALFAJ61467.AE
Buffer, AMPSO, 0,2M solution, pH 8,0
CAS No.: 102029-60-7
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colourless
Odour: Odourless
MDL No.: MFCD00067500
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR