Buffer, AMPSO, 0,2M solution, pH 9,5

Supplier: Thermo Fisher Scientific
J63354.AEEA 39.8 EUR
J63354.AE J63354.AK
Buffer, AMPSO, 0,2M solution, pH 9,5
Buffers Biological Buffers
CAS No.: 102029-60-7
Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00067500
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR