Buffer, sodium citrate, 0,5M solution, pH 6,0

Supplier: Thermo Fisher Scientific

J61815.AKEA 48.7 EUR
J61815.AK J61815.AP
Buffer, sodium citrate, 0,5M solution, pH 6,0
Buffers Biological Buffers
Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00012462
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR