Buffer, sodium citrate, 0,5M solution, pH 6,5

Supplier: Thermo Fisher Scientific
J63888.AKEA 67.2 EUR
J63888.AK J63888.AP
Buffer, sodium citrate, 0,5M solution, pH 6,5
Buffers Biological Buffers
Storage temperature: 2 to 8 °C
Physical form: liquid
Appearance: colourless
Odour: odourless
MDL No.: MFCD00012462
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR