Methylmercury hydroxide 1 M in water
|
Danger
|
Synonyms:
Methylmercury(II) hydroxide
Packaged under argon in resealable ChemSeal™ bottles.
41253.XFEA
0
EUR
41253.XF
41253.AD
41253.AB
Methylmercury hydroxide 1 M in water
Methylmercury hydroxide
Formula:
CH₄HgO MW: 232.632 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013924 EINECS: 214-667-2 UN: 2024 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...