Folic acid

Supplier: Merck

47866.EA 46 EUR
47866.
Folic acid
Folic acid
Formula: C₁₉H₁₉N₇O₆
MDL Number: MFCD00079305
CAS Number: 59-30-3
EINECS: 200-419-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR