Lansoprazole

Supplier: U.S. Pharmacopeia

1356916.EA 413 EUR
1356916.
Lansoprazole
Lansoprazole
Formula: C₁₆H₁₄F₃N₃O₂S
MDL Number: MFCD00866873
CAS Number: 103577-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR