Strychnine hemisulphate

Supplier: Roth Carl

ROTH4846.1EA 0 EUR
ROTH4846.1
Strychnine hemisulphate
Strychnine hemisulphate
Formula: C₄₂H₄₆N₄O₈S
MDL Number: MFCD00068328
CAS Number: 60-41-3
EINECS: 200-477-7
UN: 1692
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR