Strychnine hemisulphate pentahydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI EUROPE
|
Danger
|
TCIAS0094-25GEA
154
EUR
TCIAS0094-25G
Strychnine hemisulphate pentahydrate ≥97.0% (by titrimetric analysis)
Strychnine hemisulphate pentahydrate
Formula:
C₄₂H₄₆N₄O₈S·5H₂O |
MDL Number:
MFCD00068328 CAS Number: 60491-10-3 EINECS: 200-477-7 UN: 1692 ADR: 6.1,I |
![]() |
Specification Test Results
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 97.0 % (calcd.on dried substance) |
Solubility in hot Water | almost transparency |
Drying loss | 10.5 to 13.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...