Imprimer
ICNA02157403
ICNA0215740380EA 406 EUR
ICNA0215740380 ICNA0215740305 ICNA0215740325
ICNA02157403
Hydrochlorothiazide is carbonic anhydrase inhibitor.
Soluble in dilute ammonia, dilute sodium hydroxide, methanol (50 mg/ml), ethanol. Practically insoluble in water.