Hydrochlorothiazide

Supplier: U.S. Pharmacopeia

USPH1314009EA 413 EUR
USPH1314009
Hydrochlorothiazide
Hydrochlorothiazide
Formula: C₇H₈ClN₃O₄S₂
MDL Number: MFCD00051765
CAS Number: 58-93-5
EINECS: 200-403-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR