Chlorotrispyrrolidinophosphonium hexafluorophosphate ≥98%
Supplier: Thermo Scientific
Danger
Synonyms: Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphateChlorotripyrrolidinophosphonium hexafluorophosphate
H54693.14EA 373 EUR
H54693.14 H54693.06 H54693.03
Chlorotrispyrrolidinophosphonium hexafluorophosphate ≥98%
Chloro-tris-pyrrolidinophosphonium hexafluorophosphate
Formula: C₁₂H₂₄ClF₆N₃P₂
MW: 421.73
Melting Pt: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210035
CAS Number: 133894-48-1
UN: 1759
ADR: 8,II