Imprimer

4,5-Dicyano-2-(3-pyridyl)imidazole, 97%

H52235.06EA 281 EUR
ALFAH52235.06 ALFAH52235.03
4,5-Dicyano-2-(3-pyridyl)imidazole, 97%
Purity: 97%
Molecular Formula: C10H5N5
Formula Weight: 195.18
MDL No.: MFCD16658865
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR