7-(2,3-Dihydroxypropyl)theophylline
Supplier: Molekula
Synonyms:
Dyphylline
MOLE77994840-25MGEA
0
EUR
MOLE77994840-25MG
7-(2,3-Dihydroxypropyl)theophylline
7-(2,3-Dihydroxypropyl)theophylline
Formula:
C₁₀H₁₄N₄O₄ MW: 254.24 g/mol |
MDL Number:
MFCD00005759 CAS Number: 479-18-5 EINECS: 207-526-1 |