Piceatannol, Calbiochem®
Supplier: Merck Millipore Calbiochem
Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol
527948-1EA 65.9 EUR
527948-1
Piceatannol, Calbiochem®
Piceatannol
Formula: C₁₄H₁₂O₄
MDL Number: MFCD00221715
CAS Number: 10083-24-6